- Length:17.5 cm (Without Burr)
- Cat. No.:101326-502
- Supplier no.:67-1200
- Diameter:1.9 cm
The Ideal Micro-Drill™ is designed for research applications that require surgical burrs and trephines.
- Light-weight design and Rechargeable
- No special tools needed to change burrs
- Effective balancing and handling
- One year warranty
- Hard plastic case with die-cut foam for convenient storage
The drill is constructed of light-weight aluminum alloy for balance and control and is powered by a rechargeable 6 V nickel cadmium battery. Under normal operating conditions the unit will function for 8 hours between recharging.