StrataPrep Plasmid Purification Kits, Agilent Technologies
Supplier: Avantor
Certificates
About this item
The StrataPrep plasmid preparation kits provide a rapid method for purifying plasmid DNA from bacterial cultures.
- High yields of plasmid DNA: up to 20 µg from 1.5 to 3.0 ml of culture
- Accommodates all sizes of DNA from standard 2.9 kb plasmids to 45 kb cosmids
- Transforms as efficiently as cesium chloride-purified DNA
The kit utilizes a special silica-based fiber matrix technology in a microspin-column format to capture DNA in the presence of a chaotropic salt, allowing the recovery of exceptionally clean DNA.
Contains: Solution 1, 2, 3, Wash buffer, Nuclease-removal buffer, uspin cups and receptacle tubes.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:StrataPrep Plasmid Miniprep Kit
Specifications
Related Information
Frequently Bought Together