Lead(II) oxide (Litharge), granular gold- and silver-free
Catalog # 80700-091
Supplier: Anachemia Mining
CAS Number:
Specifications
- Pack type:Pail
- Size:25 kg
- Cat. No.:80700-091
- Supplier no.:029-930-11
Specifications
Specification Test Results
- Appearance granularity:Granular
- Appearance color:Yellow to orange
- Gold (Au):∼ 1 ppb
- Silver (Ag):max 0.16 ppm
Specification Test Results
About this item
:
Merck Index:13,05431
Boiling Pt:1470 °C (1013 hPa)
Melting Pt:888 °C
Density:9.5 g/cm³ (20 °C)
EINECS:215-267-0
UN:2291
ADR:6.1,III
:223.21 g/mol
:GHS08
:GHS07
:GHS09
CAS Number:1317-36-8
Formula:PbO
MDL Number:MFCD00011164