Chromium(III) acetate solution 11,2 - 11,8% Cr
Marketsource
Supplier: Strem Chemicals Inc
Chromium(III) acetate solution 11,2 - 11,8% Cr
Supplier: Strem Chemicals Inc
Marketsource
Certificates
Jump to:
About this item
- CAS Number:1066-30-4
- MDL Number:MFCD00036429
- Density:1.28
2 Options Available Below
Product Details & Documents
- CAS Number:1066-30-4
- MDL Number:MFCD00036429
- Density:1.28
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 24-0155-1000GSpecifications:MarketsourceMarketsource
- Supplier Number: 24-0155-4000GSpecifications:MarketsourceMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



