ContactUsLinkComponent
Tacky Traxx Alert!™ High Visibility Tackified Entrance Mat Frame, High-Tech Conversions
Tacky Traxx Alert!™ High Visibility Tackified Entrance Mat Frame, High-Tech Conversions
Catalog # 76221-330
undefined
Tacky Traxx Alert!™ High Visibility Tackified Entrance Mat Frame, High-Tech Conversions
Catalog # 76221-330
Supplier Number:  TTF-ALERT!
Specifications
  • Color:
    Neon Yellow
  • Dimensions:
    56x76 cm (22x30")
  • Cat. No.:
    76221-330
Specifications
About this item
Tacky Traxx Alert!™ tackified mat frame emits a bright yellow/green glow which helps reduce accidental tripping or injury.
  • High visibility
  • Glowing edges
  • Reduce trips and injuries
  • Gripper pad base
  • Reusable
New Tacky Traxx Alert!™ tackified mat frame are so bright that they appear electric, but they are not. The frame’s edges below the mat emit a bright yellow/green glow. Tacky Traxx Alert!™ frames are designed and developed to be highly visible. This new design can help reduce accidental tripping and injuries. It will also ensure that the mats are being used and will reduce contamination in controlled environments.
A complete unit consists of a reusable high visibility base frame with a tackified 30 sheet clear mat (see 76193-606) on its surface. Each numbered mat can be replenished. The frame comes with a gripper backing that allows it to be placed on virtually any surface, including carpet. The gripper backing also allows it to be relocated anytime.
Accessories information: Purchase clear Tacky Traxx along with this product (see 76193-606).
Packaging: 23×30×1", 7 lb. packaging.