ContactUsLinkComponent
Bee-Safe™ High Visibility ESD/Anti-Static Heel Grounder, Neon Yellow, High-Tech Conversions
Bee-Safe™ High Visibility ESD/Anti-Static Heel Grounder, Neon Yellow, High-Tech Conversions
Catalog # 76221-338
undefined
Bee-Safe™ High Visibility ESD/Anti-Static Heel Grounder, Neon Yellow, High-Tech Conversions
Catalog # 76221-338
Supplier Number:  GH-HS-HIVIS
Specifications
  • Color:
    Neon Yellow
  • Size:
    One Size
  • Cat. No.:
    76221-338
Specifications
About this item
These neon yellow ESD heel straps provide a visual aid to ensure all personnel are in compliance within the lab.
  • High visibility
  • ESD safe
  • One size
  • Reusable
These heel straps offer the perfect solution to those working in sensitive electronic manufacturing environments. ESD heel straps allow any electrical charge produced or transferred to dissipate and stay grounded, which will ultimately protect the electronic products. The bright color provides a visual aid to ensure all personnel are in compliance within the lab.
Packaging: 7×7×7", 0.5 lbs. packaging.