Sodium thiosulfate 0.0394 M Na2S2O3 (0.0394N) in aqueous solution volumetric solution, for the determination of iodine adsorption number (ASTM D 1510), Fluka™
Catalog # BJ-35224-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ-35224-1L
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:158.11 g/mol
Boiling Pt:100 °C (1.013 hPa)
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499