Order Entry
United States
ContactUsLinkComponent
Recombinant Leukocyte immunoglobulin-like receptor subfamily A member 5
Recombinant Leukocyte immunoglobulin-like receptor subfamily A member 5
Certificates
Recombinant Leukocyte immunoglobulin-like receptor subfamily A member 5

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Protein/Peptide Name:
    Leukocyte immunoglobulin-like receptor subfamily A member 5
  • Species:
    Human

Specifications

Frequently Bought Together