Orders LinkContactUsLinkComponent
Human Recombinant Testin
Marketsource
Human Recombinant Testin
Marketsource
Certificates
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Testin is produced by our E.coli expression system and the target gene encoding Met1-Ser421 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 102864-214
  • 102864-216
  • 102864-218
  • 102864-220
  • Protein/Peptide Name:
  • Testin
  • Testin
  • Testin
  • Testin
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Human
Specifications
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 10 μg
    Log in for availability
    Log in for availability
    Each (10µG)
    Each (10µG) - $120.83
    $120.83
    Each (10µG)
       $120.83
    Supplier Number: CE60-10UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102864-214
    Protein/Peptide Name
    Testin
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Marketsource
  • 1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $2,537.45
    $2,537.45
    Each (1mg)
       $2,537.45
    Supplier Number: CE60-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102864-216
    Protein/Peptide Name
    Testin
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Marketsource
  • 500 μg
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $1,776.22
    $1,776.22
    Each (500µG)
       $1,776.22
    Supplier Number: CE60-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102864-218
    Protein/Peptide Name
    Testin
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Marketsource
  • 50 μg
    Log in for availability
    Log in for availability
    Each (50µG)
    Each (50µG) - $363.58
    $363.58
    Each (50µG)
       $363.58
    Supplier Number: CE60-50UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102864-220
    Protein/Peptide Name
    Testin
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Marketsource