Order Entry
United States
Orders LinkContactUsLinkComponent
 

Human Recombinant Gdnf

Marketsource
Human Recombinant Gdnf
Marketsource
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products

  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels

Recombinant Human GDNF is produced by our E.coli expression system and the target gene encoding Ser78-Ile211 is expressed.

Accessories information: Accessories not included.

Delivery information: Shipped Via FedEx

Packaging: Shipped at ambient temperature via FedEx envelope.

Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 102860-614
  • 102860-620
  • 102860-618
  • 102860-616
  • Enzyme Name:
  • Gdnf
  • Gdnf
  • Gdnf
  • Gdnf
  • Enzyme type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Human

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 μg
    Log in for availability
    Log in for availability
    Each (10µG)
    Each (10µG) - $173.55
    $173.55
    Each (10µG)
       $173.55

    Supplier Number: C226-10UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102860-614
    Enzyme Name
    Gdnf
    Enzyme type
    Recombinant
    Species
    Human
    Marketsource
  • 50 μg
    Log in for availability
    Log in for availability
    Each (50µG)
    Each (50µG) - $507.49
    $507.49
    Each (50µG)
       $507.49

    Supplier Number: C226-50UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102860-620
    Enzyme Name
    Gdnf
    Enzyme type
    Recombinant
    Species
    Human
    Marketsource
  • 500 μg
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $1,776.22
    $1,776.22
    Each (500µG)
       $1,776.22

    Supplier Number: C226-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102860-618
    Enzyme Name
    Gdnf
    Enzyme type
    Recombinant
    Species
    Human
    Marketsource
  • 1 mg
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $2,537.45
    $2,537.45
    Each (1mg)
       $2,537.45

    Supplier Number: C226-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102860-616
    Enzyme Name
    Gdnf
    Enzyme type
    Recombinant
    Species
    Human
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.