Human Recombinant Beta-Nerve Growth Factor
About this item
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human pro-Nerve Growth Factor is produced by our E.coli expression system and the target gene encoding Glu19-Ala241 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
4 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human pro-Nerve Growth Factor is produced by our E.coli expression system and the target gene encoding Glu19-Ala241 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 102859-478
- 102859-484
- 102859-482
- 102859-480
- Protein/Peptide Name:
- Beta-Nerve Growth Factor
- Beta-Nerve Growth Factor
- Beta-Nerve Growth Factor
- Beta-Nerve Growth Factor
- Protein/Peptide Type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Human
- Human
- Human
- Human
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: C078-10UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102859-478Protein/Peptide NameBeta-Nerve Growth FactorProtein/Peptide TypeRecombinantSpeciesHumanMarketsource - Supplier Number: C078-50UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102859-484Protein/Peptide NameBeta-Nerve Growth FactorProtein/Peptide TypeRecombinantSpeciesHumanMarketsource - Supplier Number: C078-500UGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102859-482Protein/Peptide NameBeta-Nerve Growth FactorProtein/Peptide TypeRecombinantSpeciesHumanMarketsource - Supplier Number: C078-1MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:Marketsource
More
Specifications
Cat. No.102859-480Protein/Peptide NameBeta-Nerve Growth FactorProtein/Peptide TypeRecombinantSpeciesHumanMarketsource