Orders LinkContactUsLinkComponent
Mouse Recombinant Complement C5
Marketsource
Mouse Recombinant Complement C5
Marketsource
Mouse Recombinant Complement C5
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Complement Component C5 is produced by our E.coli expression system and the target gene encoding Asn679-Arg755 is expressed.
 : Accessories not included.
 : Shipped Via FedEx
 : Shipped at ambient temperature via FedEx envelope.
 : This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
  • Catalog No:
  • 102859-454
  • 102859-460
  • 102859-458
  • 102859-456
  • Protein/Peptide Name:
  • Complement C5
  • Complement C5
  • Complement C5
  • Complement C5
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Mouse
  • Mouse
  • Mouse
Product Family Options
  • Size
  • 10 μg
    Each (10µG)
    Each (10µG) - $155.98
    $155.98
    Each (10µG)
       $155.98
     : C075-10UG
    Marketsource
    Specifications
    Cat. No.
    102859-454
    Protein/Peptide Name
    Complement C5
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Marketsource
  • 50 μg
    Each (50µG)
    Each (50µG) - $461.35
    $461.35
    Each (50µG)
       $461.35
     : C075-50UG
    Marketsource
    Specifications
    Cat. No.
    102859-460
    Protein/Peptide Name
    Complement C5
    Protein/Peptide Type
    Recombinant
    Species
    Mouse
    Marketsource
  • 500 μg
    Each (500µG)
    Each (500µG) - $1,776.22
    $1,776.22
    Each (500µG)
       $1,776.22
     : C075-500UG
    Marketsource
    Specifications
    Cat. No.
    102859-458
    Protein/Peptide Name
    Complement C5
    Protein/Peptide Type
    Recombinant
    Species
    Mouse
    Marketsource
  • 1 mg
    Each (1mg)
    Each (1mg) - $2,537.45
    $2,537.45
    Each (1mg)
       $2,537.45
     : C075-1MG
    Marketsource
    Specifications
    Cat. No.
    102859-456
    Protein/Peptide Name
    Complement C5
    Protein/Peptide Type
    Recombinant
    Species
    Mouse
    Marketsource