Order Entry
United States
ContactUsLinkComponent
Anti-DM1 Mouse Polyclonal Antibody
Anti-DM1 Mouse Polyclonal Antibody
Catalog # 102993-810
Supplier:  Eagle Biosciences
Anti-DM1 Mouse Polyclonal Antibody
Catalog # 102993-810
Supplier:  Eagle Biosciences
Supplier Number:  30698-1ML

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antigen Symbol:
    DM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Size:
    1 ml
  • Cat. No.:
    102993-810
  • Supplier no.:
    30698-1ML

Specifications

About this item

Type:
Antigen: DM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: