ContactUsLinkComponent
Dangerous Goods UN EPS Foam Packaging, Qorpak
Dangerous Goods UN EPS Foam Packaging, Qorpak
  16465-066
Small Business Enterprise
undefined
Dangerous Goods UN EPS Foam Packaging, Qorpak
  16465-066
 :  HMS-6551M
Small Business Enterprise
  • Capacity:
    4 L
  • Description:
    UN EPS foam packaging with 4 L amber glass jug and phenolic cap
  • Cat. No.:
    16465-066
  • Un markings:
    4G/X 9.1/S
  • Supplier no.:
    HMS-6551M
EPS Foam provides better protection and cushioning than standard fibreboard packaging.
 : Meets UN 4G standards for Packing Groups II and III.
 : This package contains a pressure tested amber plastic coated glass jug and phenolic cap. For air shipments, check the regulations for any absorbent requirements and quantity limitations. Includes LDPE bag. Absorbent is sold separately.
 : Sold in a convenience pack of 4.