Order Entry
United States
ContactUsLinkComponent
 
Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered
Catalog # 16777-160
Supplier:  Cytiva
CAS Number:  
undefined
Sodium hydrogen carbonate 75 g/l in aqueous solution, HyClone™, sterile filtered
Catalog # 16777-160
Supplier:  Cytiva
Supplier Number:  SH30033.01
CAS Number:  

Specifications

  • Pack type:
    PETG bottle, square
  • Size:
    100 ml
  • Cat. No.:
    16777-160
  • Supplier no.:
    SH30033.01

Specifications

Specification Test Results

  • Endotoxin concentration:
    ≤1.0 EU/mL
  • Sterilization:
    0.1 µm sterile filtered
  • pH:
    7.9 - 9.4

Specification Test Results

About this item

 :
 :84.01 g/mol
Storage Temperature:Ambient
CAS Number:144-55-8
Formula:NaHCO₃
MDL Number:MFCD00003528