About this item
Designed for working with air-sensitive materials, these Suba Seal® closures have a turnover flange molded to grip the outside of the container neck, promoting a double seal in conjunction with the annular serrations on the hollow plug.
- Larger fitting surface
- Serrated area holds pressure better
- More solvent resistant
The membrane on top is thick and self-sealing, permitting a large number of syringe piercings. The annular serrations depress themselves against the inside wall of the bottle or flask neck, making each serration a suction sealing point.