Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Plasmid Miniprep Kit, BioVision
Plasmid Miniprep Kit, BioVision
Plasmid Miniprep Kit, BioVision
Catalog # 76211-890
Supplier:
Avantor
CAS Number:
Expand image
Plasmid Miniprep Kit, BioVision
Catalog # 76211-890
Supplier:
Avantor
CAS Number:
Specifications
Description:
Plasmid Miniprep Kit
Specifications
About this item
Product Details
Documentation
Product Details
Documentation