ContactUsLinkComponent
Chitotriosidase Activity Assay Kit (Fluorometric), BioVision
Chitotriosidase Activity Assay Kit (Fluorometric), BioVision
Catalog # 76211-464
Supplier:  Avantor
CAS Number:  
undefined
Chitotriosidase Activity Assay Kit (Fluorometric), BioVision
Catalog # 76211-464
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    Chitotriosidase Activity Assay Kit (Fluorometric)
  • Size:
    100 assays
  • Cat. No.:
    76211-464
Specifications
About this item
Kit provides a simple, specific, sensitive assay that can detect as low as 0.2 mU/ml of CHIT1.
  • Simple, rapid & convenient assay to measure Chitotriosidase activity
  • Includes Positive Control (Chitotriosidase)
Chitotriosidase (CHIT1 or chitinase-1), is a mammalian enzyme belonging to glycosyl hydrolase-18 family (EC 3.2.1.14), which can hydrolyze the β-(1, 4)-linkage between the adjacent N-acetyl glucosamine residues of chitin. CHIT1 is synthesized exclusively by activated macrophages both in normal and inflammatory conditions and, as a host defense factor, CHIT1 plays a pivotal role in infectious disease such as malaria and fungi infectious. Increased CHIT1 activity has also been found in patients suffering diseases such as bronchial asthma and atherosclerosis. CHIT1 has been used as a valuable diagnostic biomarker for monitoring the therapeutic efficacy of treatments for Gaucher’s disease or β-glucocerebrosidase deficiency. BioVision’s Chitotriosidase Activity Assay, utilizes a fluorogenic substrate that can be hydrolyzed by chitinase and a set of proprietary assay buffers that can distinguish specific CHIT1 activity from other hydrolases including Acidic Mammalian Chitinase. The kit provides a simple, specific, sensitive assay that can detect as low as 0.2 mU/ml of CHIT1 in a variety of biological samples.