ContactUsLinkComponent
Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit, BioVision
Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit, BioVision
Catalog # 76211-438
Supplier:  Avantor
CAS Number:  
undefined
Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit, BioVision
Catalog # 76211-438
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    Angiotensin II Converting Enzyme (ACE2) Inhibitor Screening Kit
  • Size:
    100 assays
  • Cat. No.:
    76211-438
Specifications
About this item
Simple assay for high-throughput screening of SIRT1 inhibitors/activators
  • Simple and reliable test to screen ACE2 inhibitors/activators
  • High-throughput compatible
  • Includes Inhibitor Control
Angiotensin II converting enzyme (ACE2, EC 3.4.17.23), a carboxypeptidase, is part of the renin-angiotensin system (RAS) that controls regulation of blood pressure by cleaving the C-terminal dipeptide of Angiotensin II to convert it into Angiotensin 1-7. It also cleaves Angiotensin I to produce Ang 1-9, of unknown function. ACE2 is a receptor of human coronaviruses, such as SARS and HCoV-NL63. It is expressed on the vascular endothelial cells of heart and kidney. The inhibitors of ACE2 could be able to regulate hypertension by changing vascular permeability. Screening for small molecule and peptide inhibitors might also help in finding treatment for coronavirus mediated infection. BioVision’s ACE2 Inhibitor Screening Kit can be used to screen for potent inhibitors of ACE2 activity, it utilizes the ability of an active ACE2 to cleave a synthetic MCA based peptide substrate to release a free fluorophore. The released MCA can be easily quantified using a fluorescence microplate reader. In the presence of an ACE2 specific inhibitor, the enzyme loses its peptidase activity which results in decrease of fluorescence intensity. This assay kit is simple and can be used to identify and characterize ACE2 inhibitors in a high-throughput format.