The Food Chemistry Set
Catalog # 470232-076
Supplier: Mega Molecules
Specifications
- Description:The Food Chemistry Set
- Cat. No.:470232-076
Specifications
About this item
Build carbohydrates, proteins and lipids!
- Durable plastic snap case included
- Activity guide included
This set includes 450 pieces and student activity guides for carbohydrates, proteins and lipids. Students construct monosaccharides and manipulate the models to illustrate the process of dehydration synthesis, build amino acids and simulate protein synthesis, or assemble and compare saturated and unsaturated fats.