Order Entry
United States
ContactUsLinkComponent
Reagent alcohol 50% (v/v) in water (denatured with methanol and isopropanol)
Flammable
Irritant
Health
Reagent alcohol 50% (v/v) in water (denatured with methanol and isopropanol)
Catalog # RC2546.50-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Flammable
Irritant
Health
Reagent alcohol 50% (v/v) in water (denatured with methanol and isopropanol)
Catalog # RC2546.50-16
Supplier:  Ricca Chemical
Supplier Number:  2546.50-16
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 ml
  • Cat. No.:
    RC2546.50-16
  • Supplier no.:
    2546.50-16

Specifications

About this item

 :
UN:1987
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
CAS Number:64-17-5
MDL Number:MFCD00003568