Manual microsyringe pump provides an economical means for any application requiring injecting/withdrawing a calibrated volume under manual control.
- Delivers precise fluid volumes
- Accommodates most syringes
- Digital micrometer option
The 51222 model can be used for infusion or withdrawal with syringes 25 µl or larger. A special micrometer with a non-rotating barrel and a clamp binds the syringe plunger to the micrometer.
The 51218 model has a rotating barrel micrometer that can only be used for infusion.
Applications includes ICSI microtitration, chromatography, microinjections.