Order Entry
United States
ContactUsLinkComponent
Triethylamine ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
Triethylamine ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
Catalog # BJ65897-50ML
CAS Number:  
undefined
Triethylamine ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
Catalog # BJ65897-50ML
Supplier Number:  65897-50ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    50 ml
  • Cat. No.:
    BJ65897-50ML

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    min. 99.5 %
  • Refractive index (n 20/D):
    1.400 - 1.402
  • Appearance:
    complying
  • Non-volatile matter:
    max. 0.003 %
  • Water (Karl Fischer):
    max. 0.5 %
  • Aluminium (Al):
    max. 0.2 ppm
  • Barium (Ba):
    max. 0.1 ppm
  • Bismuth (Bi):
    max. 0.1 ppm
  • Calcium (Ca):
    max. 0.5 ppm
  • Cadmium (Cd):
    max. 0.05 ppm
  • Cobalt (Co):
    max. 0.02 ppm
  • Chromium (Cr):
    max. 0.02 ppm
  • Copper (Cu):
    max. 0.02 ppm
  • Iron (Fe):
    max. 0.1 ppm
  • Potassium (K):
    max. 0.5 ppm
  • Lithium (Li):
    max. 0.1 ppm
  • Magnesium (Mg):
    max. 0.1 ppm
  • Manganese (Mn):
    max. 0.02 ppm
  • Molybdenum (Mo):
    max. 0.02 ppm
  • Sodium (Na):
    max. 0.5 ppm
  • Nickel (Ni):
    max. 0.02 ppm
  • Lead (Pb):
    max. 0.1 ppm
  • Strontium (Sr):
    max. 0.1 ppm
  • Zinc (Zn):
    max. 0.1 ppm
  • UV- cutoff:
    250.0 - 278.0 nm

Specification Test Results

About this item

 :
 :101.19 g/mol
Boiling Pt:90 °C
Melting Pt:–115 °C
Density:0.73 g/cm3 (20 °C)
Flash Pt:–11 °C
Storage Temperature:Ambient
EINECS:204-469-4
UN:1296
ADR:3 (8),II
 :GHS02
 :GHS05
 :GHS07
CAS Number:121-44-8
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051