Sucrose Solutions for Refractometers, Atago
Supplier: Atago
Certificates
About this item
Sucrose solutions are used for confirming the Brix measurements of refractometers.
- Standard solutions have a shelf life of 6 weeks
- High accuracy solutions have a shelf life of 10 days
Frequently Bought Together