GemClean Cross-Flow II Nebulizer, PerkinElmer
Complete chemically inert Cross-Flow II nebulizer assembly for ICP-MS. Specially processed to reduce BEC levels below 1 ppb.
- Suitable for NexION/ELAN DRCe/9000/6X00/5000/500/250
Contains an end cap with a GemTip clear sapphire sample tip and a GemTip red ruby argon tip optimized for ICP-MS.
Frequently Bought Together