Single Stage Pneumatic Gas Regulators, PerkinElmer
Catalog # 10138-308
Supplier: PerkinElmer
Specifications
- Description:Single-stage pneumatic gas regulator
- Cat. No.:10138-308
- Inlet pressure:3000 psi (Max.)
- Supplier no.:N2410173
Specifications
About this item
The single-stage brass body regulator with neoprene diaphragm is appropriate for nitrogen gas, air, or argon.
- Used for driving the pneumatic valves of the PerkinElmer 2400 and PerkinElmer 2410
- For direct connection to the ¹/₈" gas line, order the Swagelok ¹/₄ to ¹/₈" reducing fitting (P/N 09903471)
Delivery: Includes a 590 to 580 adapter.