Order Entry
United States
ContactUsLinkComponent
(Z)-4-Hydroxytamoxifen, Sigma-Aldrich®
(Z)-4-Hydroxytamoxifen, Sigma-Aldrich®
Catalog # 80057-566
Supplier:  Avantor
CAS Number:  
(Z)-4-Hydroxytamoxifen, Sigma-Aldrich®
Catalog # 80057-566
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    5 mg
  • Cat. No.:
    80057-566
  • Supplier no.:
    579002-5MG

Specifications

About this item

 :
 :387.51 g/mol
CAS Number:68047-06-3
Formula:C₂₆H₂₉NO₂
MDL Number:MFCD00278780