ContactUsLinkComponent
Zearalenon Immunaffinity Column, Eagle Biosciences, Inc.
Zearalenon Immunaffinity Column, Eagle Biosciences, Inc.
Catalog # 103388-994
Supplier:  Eagle Biosciences
Zearalenon Immunaffinity Column, Eagle Biosciences, Inc.
Catalog # 103388-994
Supplier:  Eagle Biosciences
Supplier Number:  BTZE315005
Specifications
  • Description:
    Zearalenon Immunaffinity Column, 10 columns
  • Cat. No.:
    103388-994
Specifications
About this item
The Zearalenon immunoaffinity column provides the Zearalenon (ZON) determination in food and feed.
  • Sensitivity: 0.2 to 20 ng
  • Dynamic range: 0.2 to 20 ng
  • Sample size: 10 columns
This instruction of Zearalenon (ZON) determination in food and feed focuses on the enrichment step of extract using immunoaffinity column (IAC) and quantification with HPLC. Accepted laboratory extraction methods could be maintained. Full performance of the IAC column is given if pronounced criteria regarding organic solvent tolerance, elution process of analyte and working range of column is followed. Many pretreatment methods of Zearalenon determination in food and feed show low sensitivity because of interfering substances if problematic matrices are applied. This method of content determination of Zearalenon combines the high selectivity of an immunoaffinity column (IAC) with its potential to concentrate elute and additional step of purification by HPLC column. For the commodity extraction step a literature method is given. The given apparatus (e.g. HPLC system) might serve as example among other possibilities.