ContactUsLinkComponent
Aflatoxin, Ochratoxin, and Zearalenon Immunoaffinity Column , Eagle Biosciences, Inc.
Aflatoxin, Ochratoxin, and Zearalenon Immunoaffinity Column , Eagle Biosciences, Inc.
Catalog # 103388-980
Supplier:  Eagle Biosciences
Aflatoxin, Ochratoxin, and Zearalenon Immunoaffinity Column , Eagle Biosciences, Inc.
Catalog # 103388-980
Supplier:  Eagle Biosciences
Supplier Number:  BTCZ322005
Specifications
  • Description:
    Aflatoxin, Ochratoxin, and Zearalenon Immunoaffinity Column, 10 columns
  • Cat. No.:
    103388-980
Specifications
About this item
Aflatoxin, Ochratoxin and Zearalenon (AOZ) in food and feed focuses using immunoaffinity column.
  • Sensitivity: 0.02 to 3 ng AFL, 0.02 to 2 ng OTA, 0.2 to 20 ng ZON
  • Dynamic Range: 0.02 to 3 ng AFL, 0.02 to 2 ng OTA, 0.2 to 20 ng ZON
  • Sample Size: 10 columns
This instruction of simultaneous Aflatoxin, Ochratoxin and Zearalenon (AOZ) determination in food and feed focuses on the enrichment step of extract using immunoaffinity column (IAC) and quantification with HPLC. Accepted laboratory extraction methods could be maintained. Full performance of the IAC column is given if pronounced criteria regarding organic solvent tolerance, elution process of analyte and working range of column is followed. Many pretreatment methods of Aflatoxin, Ochratoxin and Zearalenon determination in food and feed show low sensitivity because of interfering substances if problematic matrices are applied. This method of content determination of Aflatoxins, Ochratoxins and Zearalenon combines the high selectivity of an immunoaffinity column (IAC) with its potential to concentrate eluate and additional step of purification by HPLC column. Please notice that this instruction focuses on the handling with the IAC column. For the commodity extraction step a literature method is given. The given apparatus (e.g. HPLC system) might serve as example among other possibilities.