Order Entry
United States
ContactUsLinkComponent
Zonisamide HPLC Assay, Eagle Biosciences, Inc.
Zonisamide HPLC Assay, Eagle Biosciences, Inc.
Catalog # 102995-118
Supplier:  Eagle Biosciences
CAS Number:  
Zonisamide HPLC Assay, Eagle Biosciences, Inc.
Catalog # 102995-118
Supplier:  Eagle Biosciences
Supplier Number:  ZON31-H100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Zonisamide HPLC Assay
  • Size:
    1 kit
  • Cat. No.:
    102995-118
  • Supplier no.:
    ZON31-H100

Specifications

About this item

The Zonisamide HPLC Assay detects Zonisamide in serum and plasma.

  • Incubation Time: 15 minutes
  • Sample Size: 100 µl
  • Sample Type: Serum, Plasma

The antiepileptic drug Zonisamide is used as add on therapy for the treatment of epileptic disorders. Zonisamide, a benzisoxazol-derivative, doesn´t show any similarities to other antiepileptic drugs on the market. The therapeutic range of the serum concentration of this drug during dosing is rather narrow; high doses often cause side effects such as ataxia, nystagumus, diplopia, drowsiness, loss of appetite and more. Clinically significant effects are observed with plasma levels higher than 16.5 mg/l, and some toxic symptoms may occur when the plasma concentration exceeds 40 mg/l (Sackellares et al, 1985). Therefore the determination of the plasma concentration is required in epileptic patients, in order to establish a proper concentration of the drug for the inhibition of epileptic seizures and to avoid side effects during chronic drug administration.