Order Entry
United States
ContactUsLinkComponent
Vitamin C HPLC Assay, Eagle Biosciences
Vitamin C HPLC Assay, Eagle Biosciences
Catalog # 102995-104
Supplier:  Eagle Biosciences
CAS Number:  
Vitamin C HPLC Assay, Eagle Biosciences
Catalog # 102995-104
Supplier:  Eagle Biosciences
Supplier Number:  VIC31-H100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Vitamin C HPLC Assay
  • Size:
    1 kit
  • Cat. No.:
    102995-104
  • Supplier no.:
    VIC31-H100

Specifications

About this item

The Vitamin C HPLC Assay is intended to detect vitamin C in Li-heparinate plasma and serum.

  • Incubation Time: 10 minutes
  • Sample Size: 200 µl
  • Sample Type: Lithium Heparinate Plasma

Vitamin C is a strong reducing agent. The oxidation of vitamin C leads to dehydroascorbic acid via a radicalic intermediate, in vivo. The three forms constitute a reversible redox-system. The most important function of ascorbic acid is the catalization in the biosynthesis of collagen. It is important for the de novo synthesis of bone, cartilage and teeth and wound healing. Vitamin C is needed for the synthesis of noradrenalin. Vitamin C plays an important role as antioxidant. It protects other molecules and cell structures from free radicals and reactive oxygen species. Ascorbic acid promotes the resorption of iron in the intestine and reduces the production of nitrosamines which might be carcinogenic.