Order Entry
United States
ContactUsLinkComponent
Vitamin B6 Whole Blood HPLC Assay, Eagle Biosciences, Inc.
Vitamin B6 Whole Blood HPLC Assay, Eagle Biosciences, Inc.
Catalog # 102995-090
Supplier:  Eagle Biosciences
CAS Number:  
Vitamin B6 Whole Blood HPLC Assay, Eagle Biosciences, Inc.
Catalog # 102995-090
Supplier:  Eagle Biosciences
Supplier Number:  VB630-H100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Vitamin B6 Whole Blood HPLC Assay
  • Size:
    1 kit
  • Cat. No.:
    102995-090
  • Supplier no.:
    VB630-H100

Specifications

About this item

The Vitamin B6 HPLC Assay Kit is intended to detect vitamin B6 in EDTA-blood.

  • Incubation Time: 30 minutes
  • Sample Size: 100 µl
  • Sample Type: EDTA Whole Blood

The vitamins pyridoxin, pyridoxal and pyridoxamin and the appropriate phosphate products are summarized as vitamin B6. All forms can be transformed into the active form pyridoxal-5-phosphate. Vitamin B6 functions as a coenzyme and is essential for more than 50 reactions in the protein, carbohydrate and fatty acid metabolism thereby synthesizing, transforming or degrading amino acids. In protein metabolism vitamin B6 supports the resorption of amino acids and their transport into the cells. Furthermore vitamin B6 contributes to the synthesis of neurotransmitters and amine products (histamine).