- Description:Phenylalanine, Tyrosine & Tryptophan HPLC Assay
- Size:1 kit
- Cat. No.:102994-886
- Supplier no.:PNL31-H100
The Phenylalanine Tyrosine & Tryptophan HPLC Assay detects phenylalanine, tyrosine and tryptophan.
- Incubation Time: 15 minutes
- Sample Size: 50 µl
- Sample Type: plasma, whole blood, filter spots
The Eagle Biosciences Phenylalanine, Tyrosine & Tryptophan HPLC Assay kit makes it possible to determine phenylalanine, tyrosine and tryptophan in an easy, fast and precise method. The Phenylalanine, Tyrosine & Tryptophan HPLC Assay kit includes all reagents for preparation and separation of the samples with exception of the columns (IC4000rp) and the controls (IC4000ko). Both can be supplied by Eagle Biosciences. Besides the complete test kits it is possible to order all components separately. Please request our single component price list.