ContactUsLinkComponent
25-OH Vitamin D3 HPLC Column, Eagle Biosciences, Inc.
  102994-532
25-OH Vitamin D3 HPLC Column, Eagle Biosciences, Inc.
  102994-532
 :  IC3401RP
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    25-OH Vitamin D3 HPLC Column, 1 column
  • Cat. No.:
    102994-532
  • Supplier no.:
    IC3401RP
25-OH Vitamin D3 HPLC Column can be used with the HPLC Kit and can be used for 500 to 1000 samples.
  • For use with 25-OH Vitamin D3 HPLC assay kit
The Eagle Biosciences 25-OH Vitamin D3 HPLC Assay kit makes it possible to determine the vitamin in an easy, fast and precise method. The kit includes all reagents in ready to use form for preparation and separation of the samples with exception of the column (IC3401rp) and the controls (IC3401ko). Vitamin D is one of the most important essential bioregulators of the Ca²⁺⁻ and phosphate metabolism in higher animals. Together with the peptide hormones parathormone and calcitonin it is responsible to obtain the Ca²⁺⁻ and phosphate homoeostasis. The most important physiological functions of vitamin D are the stimulation of the intestinal resorption of Ca²⁺ and phosphate and their incorporation in the bone matrix.