ContactUsLinkComponent
Malondialdehyde (MDA) HPLC Column, Eagle Biosciences, Inc.
Malondialdehyde (MDA) HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-492
Supplier:  Eagle Biosciences
Malondialdehyde (MDA) HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-492
Supplier:  Eagle Biosciences
Supplier Number:  IC1900RP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Malondialdehyde (MDA) HPLC Column, 1 column
  • Environmentally Preferable:
  • Cat. No.:
    102994-492
  • Supplier no.:
    IC1900RP
Specifications
About this item
Malondialdehyde (MDA) HPLC assay kit column can be used for about 500 to 1000 samples.
  • For use with Malondialdehyde (MDA) HPLC assay kit
In a healthy body, oxidative and reductive processes are in a balance. Free radicals (reactive oxygen species) are eliminated by antioxidants. In case of a lack of antioxidants free radicals react with cell structures. A reaction of free radicals with unsaturated fatty acids leads to lipid peroxidation products. A reaction of free radicals with poly-unsaturated fatty acids generates malondialdehyde and/or 4-hydroxynonenal. These secondary lipid peroxidation products might react with other molecules in the cell. Modifications of the DNA-based adenine or guanine result in incorrect transcripts. A reaction of free radicals with proteins leads to an alteration or loss of function. The creation of neoAntigenss is possible. NeoAntigenss are recognized by the immune system, thus resulting in autoimmune diseases. The participation of lipid peroxidation products have been discussed in several diseases such as atherosclerosis, tumor genesis, rheumatism and reperfusion injury after transplantation.