ContactUsLinkComponent
Ubiquinone / Coenzyme Q10 HPLC Column, Eagle Biosciences, Inc.
Ubiquinone / Coenzyme Q10 HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-484
Supplier:  Eagle Biosciences
Ubiquinone / Coenzyme Q10 HPLC Column, Eagle Biosciences, Inc.
Catalog # 102994-484
Supplier:  Eagle Biosciences
Supplier Number:  IC1700RP
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Ubiquinone / Coenzyme Q10 HPLC Column, 1 column
  • Cat. No.:
    102994-484
  • Supplier no.:
    IC1700RP
Specifications
About this item
The Ubiquinone/Coenzyme Q10 column can be utilized for approximately 500 to 1000 samples.
  • For use with Ubiquinone / Coenzyme Q10 HPLC assay kit
Ubiquinone is a coenzyme of every cell in the body. Ubiquinone is made up by a chinonic ring and an isoprenic side chain. In humans ubiquinone can be synthesized and taken up by nutrition.