Order Entry
United States
ContactUsLinkComponent
Complement Alternative pathway ELISA, Eagle Biosciences, Inc.
Complement Alternative pathway ELISA, Eagle Biosciences, Inc.
Catalog # 102994-104
Supplier:  Eagle Biosciences
CAS Number:  
Complement Alternative pathway ELISA, Eagle Biosciences, Inc.
Catalog # 102994-104
Supplier:  Eagle Biosciences
Supplier Number:  COMPLAP330
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Complement Alternative pathway ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-104
  • Supplier no.:
    COMPLAP330

Specifications

About this item

The Eagle Biosciences Human Complement Alternative Pathway ELISA Assay kit is for Research Use Only.

  • Sensitivity: Cut-off
  • Dynamic Range: Cut-off
  • Incubation Time: 2 hours

In vitro activation of the complement sequence leads to the consumption of complement components which, in turn, leads to a decrease in their concentration. Thus, the determination of complement proteins or complement activity is used to indicate whether the complement system has been activated by an immunologic and/or pathogenic mechanism. Both functional and immunochemical complement measurements are used to evaluate subjects when a complement-activating disease is suspected or an inherited deficiency is possible. The level of complement activity evaluated by functional assays such as Eagle Biosciences Complement Alternative Pathway ELISA Assay Kit  takes into account the rate of synthesis, degradation, and consumption of the components and provides a measure of the integrity of the pathways as opposed to immunochemical methods which specifically measure the concentration of various complement components.