ContactUsLinkComponent
Folic Acid Immunoaffinity Column, Eagle Biosciences, Inc.
Folic Acid Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-018
Supplier:  Eagle Biosciences
Folic Acid Immunoaffinity Column, Eagle Biosciences, Inc.
Catalog # 102994-018
Supplier:  Eagle Biosciences
Supplier Number:  BTFS319005
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Folic Acid Immunoaffinity Column, 10 columns
  • Cat. No.:
    102994-018
  • Supplier no.:
    BTFS319005
Specifications
About this item
This method for Folic Acid combines IAC with its concentrate elute by purification with HPLC.
  • Sensitivity: 0.03 μg
  • Dynamic range: 0.03 to 5 μg
  • Sample size: 10 columns
This method of determination of Folic Acid combines the high selectivity of immunoaffinity columns (compared to other separation materials (e.g. SAX)) with its potential to concentrate elute through use of purification by a HPLC column.