Oakton® pH Testr™ 50S Spear-Tip Waterproof Pocket Tester, Premium 50 Series, Cole-Parmer
76208-090
- Calibration:Up to 3 points
- Temperature Range:0 – 60 °C (32 – 140 °F)
- Description:Spear Tip Waterproof Pocket pH Tester
- Temperature Resolution:0.1 °C (0.1 °F)
- Battery Life:250 hrs.
- Automatic Buffer Recognition:USA 4.01, 7.00, 10.01 and NIST 4.01, 6.86, 9.18
- Temperature Compensation:Manual
- pH Accuracy:±0.01 pH
- pH Range:–1 to 15
- pH Resolution:0.01 pH
- Battery Type:Four AAA 1.5 V
- Cat. No.:76208-090
Large electrode junction makes clogging virtually impossible.
- Rugged spear tip makes it ideal for soft solids and semisolid materials
- Easy-to-navigate interface for quick setup and calibration
- Replaceable, double-junction electrode sensors save time and money
- Waterproof
- LCD display
Automatic temperature compensation ensures accurate reading in varying environments.
: Includes protective plastic storage case, lanyard, and 4 AAA batteries.