ContactUsLinkComponent
HardyCHROM™ MRSA Chromogenic Medium, Hardy Diagnostics
HardyCHROM™ MRSA Chromogenic Medium, Hardy Diagnostics
Catalog # 76210-510
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyCHROM™ MRSA Chromogenic Medium, Hardy Diagnostics
Catalog # 76210-510
Supplier:  Hardy Diagnostics
Supplier Number:  P14
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Size:
    15×65 mm
  • Cat. No.:
    76210-510
Specifications
About this item
HardyCHROM™ MRSA, Contact Plate is a chromogenic medium recommended for use in the cultivation of methicillin resistant Staphylococcus aureus (MRSA) from environmental surfaces to aid in the prevention and control of MRSA infections in health care settings.
  • Read-out at 24 hours
  • Distinct color change read-out
  • Bright color development
  • Friction lid design keeps lids in place
  • Lok-Tight™ Locking Lids - optional locking feature means lids stay secured during sampling and transport
  • Molded Grid - Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
  • Certificate of Analysis available online for each lot
Each contact plate has a specified grid molded into the bottom of the plate for enumeration of microbial colonies growing on a variety of surfaces.
Caution: HardyCHROM™ MRSA, Contact Plate is not intended to diagnose MRSA infection nor to guide or monitor therapy.