HardyDisks™ Meropenem/Vaborbactam, AST, Hardy Diagnostics
Supplier: Hardy Diagnostics
Certificates
About this item
Meropenem and Vaborbactam are used for susceptibility testing. It made of impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
- Complete line of all commonly used antibiotics
- Compatible with the BBL dispensers
- Improved cartridge design helps to prevent jamming and tearing
- Cartridges are packaged in an all new canister that features a desiccant built into the lid
- Spring loaded in cartridges of 50 disks for smooth and reliable dispensing
- The last disk in each cartridge is clearly marked with dark lines around its edge to signify the need for refill
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Accessories information: Accessories available: Mueller Hinton Agar, McFarland Standard, calipers, and much more.
Frequently Bought Together