Thermal Transfer Labels, White, HCL Labels
Catalog # 76204-584
Supplier: HCL Label
Specifications
- Color:White
- Description:White thermal transfer label
- Dimensions:5.5×3.25"
- Cat. No.:76204-584
- Packaging:1000 labels/roll, 1 roll
Specifications
About this item
These thermal transfer labels are easy-to-use and are a great value.
- For use with thermal printers
- 1000 labels per roll
With a white background, these thermal transfer labels are ideal for use in any work environment.