- Length:6"
- Material:Stainless steel
- Curvature:Straight
- Cat. No.:76192-644
- Tip style:Serrated
Econo™ Virtus Splinter Forceps are commonly used in to remove larger foreign objects.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Econo™ Virtus Splinter Forceps feature a triangle tip and a single spring pliers handle. They may also be known as Ralks forceps. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.