Specifications
- Length:7"
- Material:Stainless steel
- Curvature:Straight
- Cat. No.:76192-612
Specifications
About this item
Econo™ Tuttle Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
- For more than 120 years, Sklar has set the standard for surgical instrumentation
- With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
- All Sklar products are industry standard
Econo™ Tuttle Forceps have long shafts which make them useful for working in areas such as the intestines. These forceps may also be known as Singley forceps. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.