Order Entry
United States
Orders LinkContactUsLinkComponent
Ampicillin solution 50 mg/ml

Ampicillin solution 50 mg/ml

25 ml
Catalog # 100220-262
Supplier:   Teknova
Supplier #:  A9525
Ampicillin solution 50 mg/ml
Catalog # 100220-262
Supplier:  Teknova
CAS Number:  69-53-4
Ampicillin solution 50 mg/ml

25 ml

CAS Number:  69-53-4
Price
$52.84
Log in for availability
Each (25ml)
Minimum order quantity is 25
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ampicillin is used as a selectable marker in many commonly used plasmids. A functional Ampicillin gene will confer resistance to cells containing the plasmid.

  • Inhibits cell wall synthesis by inhibiting formation of the peptidoglycan cross-linkages
  • Acts as a competitive inhibitor of transpeptidase

Inhibits bacterial cell wall synthesis by binding to one or more of the penicillin binding proteins (PBPs) that inhibit the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls, thus inhibiting cell wall biosynthesis.

  • CAS Number:69-53-4
  • Formula:C₁₆H₁₉N₃O₄S
  • MDL Number:MFCD00005175
  • Molecular Weight:349.4 g/mol
Specifications

Specifications

  • Size:
    25 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.