ContactUsLinkComponent
Helix Elite Inactivated Standard, Microbiologics
Helix Elite Inactivated Standard, Microbiologics
Catalog # 76199-858
Supplier:  Avantor
CAS Number:  
undefined
Helix Elite Inactivated Standard, Microbiologics
Catalog # 76199-858
Supplier:  Avantor
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Inactivated Enterovirus D68
  • Cat. No.:
    76199-858
Specifications
About this item
Inactivated Helix Elite Molecular Standards are used for every step of the molecular testing process, from extraction, to amplification and detection.
  • Fully intact, non-viable process controls
  • FDA listed and CE Marked as an In-Vitro Diagnostic (IVD) Medical Device
  • Used for every step of the molecular testing process, from extraction, to amplification and detection
  • Quick dissolving lyophilized pellets may be reconstituted in buffer or transport medium
  • Processed using the same protocols as the patient sample
Inactivated Helix Elite Molecular Standards are used for every step of the molecular testing process, from extraction, to amplification and detection. Quick dissolving lyophilized pellets may be reconstituted in buffer or transport medium. These are FDA listed and CE Marked as an In-Vitro Diagnostic (IVD) Medical Device, processed using the same protocols as the patient sample, and fully intact, non-viable process controls.