ContactUsLinkComponent
Fixed Angle Rotor, Eppendorf
Fixed Angle Rotor, Eppendorf
Catalog # 20901-055
Supplier:  Eppendorf
Product available on GSA Advantage®
undefined
Fixed Angle Rotor, Eppendorf
Catalog # 20901-055
Supplier:  Eppendorf
Supplier Number:  022366726
Product available on GSA Advantage®
Specifications
  • Description:
    18-Well Fixed-Angle Rotor with Cover
  • Cat. No.:
    20901-055
  • No. of places:
    18
  • For use with:
    Eppendorf Model 5415C centrifuge
  • Tube capacity:
    1.5 / 2.0 ml
  • Supplier no.:
    022366726
Specifications
About this item
Rotor for Microcentrifuge Model 5415C.