ContactUsLinkComponent
RMESFect™ CRISPR Transfection Reagent, OZ Biosciences
RMESFect™ CRISPR Transfection Reagent, OZ Biosciences
Catalog # 103537-334
Supplier:  OZ Biosciences
CAS Number:  
RMESFect™ CRISPR Transfection Reagent, OZ Biosciences
Catalog # 103537-334
Supplier:  OZ Biosciences
Supplier Number:  RMC70500
CAS Number:  
Specifications
  • Description:
    RMESFect™ CRISPR Transfection Reagent
  • Size:
    500 µL
  • Cat. No.:
    103537-334
Specifications
About this item
RmesFect™ CRISPR transfection Reagent based on the TEE-technology is specifically designed for mRNA/gRNA transfection with high efficiency and low toxicity. RmesFect™ is efficient in a large variety of cells.
  • Highly efficient: achieve RNAs delivery in all cells
  • Ready-to-use: no need for additional buffer
  • Low nucleic acid amount: minimized toxicity
  • Protects RNAs against degradation
  • Compatible with any culture medium
  • Serum compatible: Medium changed not required
mRNA transfection provides several advantages over plasmid DNA delivery. No need for nuclear uptake. mRNA translation into proteins occurs in the cytoplasm. It gives faster protein expression than DNA transfection and has no genomic integration. Protein expression in a total promoter-independent manner.