ContactUsLinkComponent
SAVIEW® PLUS AP Reagent, Enzo Life Sciences
SAVIEW® PLUS AP Reagent, Enzo Life Sciences
Catalog # 76196-686
Supplier:  Enzo Life Sciences
CAS Number:  
SAVIEW® PLUS AP Reagent, Enzo Life Sciences
Catalog # 76196-686
Supplier:  Enzo Life Sciences
Supplier Number:  ACC111-0150
CAS Number:  
Specifications
  • Size:
    150 tests
  • Cat. No.:
    76196-686
Specifications
About this item
High sensitivity, low background, streptavidin-based nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
  • Ready-to-use reagent
  • Streptavidin-based detection with reduced carbohydrate and biotin background
  • Suitable for both manual and automated platforms
  • Streptavidin Blocker/Diluent further reduces background (sold separately)
  • 5 – 10X more sensitive than avidin based detection
  • Customize with HIGHDEF® chromogens of your choice (sold separately)
This reagent is a streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded (FFPE) tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, this reagent can be used for in-situ hybridization detection of biotinylated DNA/RNA probes.