ContactUsLinkComponent
Sterile SatPax® 1000 Pre-Wetted Cellulose/Polyester Nonwoven Cleanroom Wipes, Berkshire
Sterile SatPax® 1000 Pre-Wetted Cellulose/Polyester Nonwoven Cleanroom Wipes, Berkshire
  76204-772
 :  Berkshire
undefined
Sterile SatPax® 1000 Pre-Wetted Cellulose/Polyester Nonwoven Cleanroom Wipes, Berkshire
  76204-772
 :  Berkshire
 :  SSP100000124
  • Description:
    Sterile SatPax® 1000 Wipes, C-Fold
  • Environmentally Preferable:
  • Cat. No.:
    76204-772
  • Packaging:
    30 Wipes/Pack, 24 Packs/Case (Stacked)
  • W×L:
    22.9×22.9 cm (9×9)"
Gamma irradiated and sterile validated to a 10–6 Sterility Assurance Level per AAMI guidelines.
  • Low extractables and fiber and particle counts
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • High saturation level is ideal for removing cleaning and disinfecting residues in sterile environments
Sterile SatPax® 1000 combines Durx® 670 nonwoven wipers composed of a hydroentangled nonwoven blend of 55% cellulose and 45% polyester with a high saturation level of 70% IPA and 30% DI water. This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles, and inconsistent wetting and cleaning associated with wetting a dry wiper. Making the product ideal for cleaning critical sterile processing environments.
 : Designed for use in ISO class 5 and higher cleanroom environments. Cleanroom Class 100 – 100,000.